Micro soxhlet extraction apparatus, WHEATON®
Soxhlet ekstraktor, — ml
Katalog # WHEA415502
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifikationer
- Beskrivelse:Soxhlet ekstraktor
- Seq:03
- Kolbes volumen:— ml