Orders LinkContactUsLinkComponent
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Mikro soxhlet-ekstraktion, komplet, 25 ml
Katalog # WHEA415500
Leverandør:   DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Katalog # WHEA415500
Leverandør:  DWK Life Sciences
Micro soxhlet extraction apparatus, WHEATON®
Micro soxhlet extraction apparatus, WHEATON®
Mikro soxhlet-ekstraktion, komplet, 25 ml
Price
8.370,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifikationer
  • Beskrivelse:
    Mikro soxhlet-ekstraktion, komplet
  • Seq:
    01
  • Kolbes volumen:
    25 ml