Micro soxhlet extraction apparatus, WHEATON®
Mikro soxhlet-ekstraktion, komplet, 25 ml
Katalog # WHEA415500
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifikationer
- Beskrivelse:Mikro soxhlet-ekstraktion, komplet
- Seq:01
- Kolbes volumen:25 ml