Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cover glasses

Cover glasses

Cover glasses, 0,13 - 0,16 mm, 15 mm, Glas
Catalog # BELC1943-10015A
Marketsource
Supplier:   BELLCO GLASS
Cover glasses
Catalog # BELC1943-10015A
Supplier:  BELLCO GLASS
Marketsource
Cover glasses
Cover glasses

Cover glasses, 0,13 - 0,16 mm, 15 mm, Glas

Price
196,17 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Round cover slips made of glass.

Specifications
  • Ø:
    15 mm
  • Beskrivelse:
    Cover glasses
  • Materiale:
    Glas
  • Seq:
    004
  • Tykkelse:
    0,13 - 0,16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.