Orders LinkContactUsLinkComponent
Sprøjtedispenser, Microlab® 600
Sprøjtedispenser, Microlab® 600
Dual syringe pump, Microlab 635, two 1 ml BFP default syringes
Katalog # 613-1569
Leverandør:   HAMILTON BONADUZ
Sprøjtedispenser, Microlab® 600
Katalog # 613-1569
Leverandør:  HAMILTON BONADUZ
Sprøjtedispenser, Microlab® 600
Sprøjtedispenser, Microlab® 600
Dual syringe pump, Microlab 635, two 1 ml BFP default syringes
Price
56.750,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
  • Dispenserer volumener fra 100 nl til 50 ml
  • Stabil væskevej, der kan anvendes til barske kemikalier
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
Certifications: CE marked. CSA certified.
Accessories information: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Ordering information: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
Specifikationer
  • Beskrivelse:
    Dual syringe pump, Microlab 635, two 1 ml BFP default syringes
  • Seq:
    08
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Luer lock conversion kit
    Luer lock conversion kit
    HAMI10150621
    Log ind for tilgængelighed
    1 * 1 Items - 996,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    HAMI1171-02
    Log ind for tilgængelighed
    1 * 1 Items - 648,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    HAMI1173-02
    Log ind for tilgængelighed
    1 * 1 Items - 648,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Screen protector, PET
    Screen protector, PET
    HAMI64241-01
    Log ind for tilgængelighed
    1 * 2 Items - 498,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Universal power supply, 110 - 220 VAC
    Universal power supply, 110 - 220 VAC
    HAMI61092-01
    Log ind for tilgængelighed
    1 * 1 Items - 1.100,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Cable management system for disposable tip hand probe
    Cable management system for disposable tip hand probe
    HAMI65160-01
    Log ind for tilgængelighed
    1 * 1 Items - 1.390,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Airshield for fresh air inside the instrument
    Airshield for fresh air inside the instrument
    HAMI68562-01
    Log ind for tilgængelighed
    1 * 1 Items - 3.840,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Håndprobe med dobbelt trykknap
    Håndprobe med dobbelt trykknap
    613-1624
    Log ind for tilgængelighed
    1 * 1 Items - 7.300,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    Fill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP
    HAMI1172-02
    Log ind for tilgængelighed
    1 * 1 Items - 532,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    Fill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP
    HAMI1174-02
    Log ind for tilgængelighed
    1 * 1 Items - 532,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Continuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEP
    Continuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEP
    HAMI61491-01
    Log ind for tilgængelighed
    1 * 1 Items - 805,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • New Microlab 600 controller, includes SD card slot, serial port, large and small USB port
    New Microlab 600 controller, includes SD card slot, serial port, large and small USB port
    HAMI61500-05
    Log ind for tilgængelighed
    1 * 1 Items - 24.320,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Continuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEP
    Continuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEP
    HAMI61491-02
    Log ind for tilgængelighed
    1 * 1 Items - 714,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Printer kit includes printer, power source, adapter and cable
    Printer kit includes printer, power source, adapter and cable
    613-5822
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Disposable tips for 613-1625, 5 ml
    Disposable tips for 613-1625, 5 ml
    HAMI75702
    Log ind for tilgængelighed
    1 * 250 Items - 855,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fodkontakt til ML 600-serien
    Fodkontakt til ML 600-serien
    613-1626
    Log ind for tilgængelighed
    1 * 1 Items - 1.970,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Valve plug
    Valve plug
    HAMI61729-01
    Log ind for tilgængelighed
    1 * 1 Items - 183,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Håndprobe med engangsspids, stort volumen (1 - 5 ml)
    Håndprobe med engangsspids, stort volumen (1 - 5 ml)
    613-1625
    Log ind for tilgængelighed
    1 * 1 Items - 13.810,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Probe holder and tubing stand
    Probe holder and tubing stand
    HAMI61710-01
    Log ind for tilgængelighed
    1 * 1 Items - 1.350,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEP
    Fill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEP
    613-1582
    Log ind for tilgængelighed
    1 * 1 Items - 391,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Fill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEP
    Fill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEP
    613-1583
    Log ind for tilgængelighed
    1 * 1 Items - 459,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)
    Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)
    HAMI61500-03
    Log ind for tilgængelighed
    1 * 1 Items - 2.590,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Concorde CT håndprobe
    Concorde CT håndprobe
    613-1621
    Log ind for tilgængelighed
    1 * 1 Items - 2.440,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Valve cross tube assembly
    Valve cross tube assembly
    HAMI61498-01
    Log ind for tilgængelighed
    1 * 1 Items - 341,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Venstremontering
    Venstremontering
    613-1620
    Log ind for tilgængelighed
    1 * 1 Items - 2.950,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEP
    Dispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEP
    613-1584
    Log ind for tilgængelighed
    1 * 1 Items - 399,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP
    Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP
    613-1585
    Log ind for tilgængelighed
    1 * 1 Items - 488,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Højremontering
    Højremontering
    613-1619
    Log ind for tilgængelighed
    1 * 1 Items - 2.950,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Disposable tip hand probe, 0,5 - 1000 µl
    Disposable tip hand probe, 0,5 - 1000 µl
    613-5823
    Log ind for tilgængelighed
    1 * 1 Items - 6.620,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Tubing clips
    Tubing clips
    HAMI88990
    Log ind for tilgængelighed
    1 * 5 Items - 523,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Basic to Advanced Controller upgrade kit, including 2 GB SD card
    Basic to Advanced Controller upgrade kit, including 2 GB SD card
    HAMI61500-02
    Log ind for tilgængelighed
    1 * 1 Items - 6.440,00 kr