Sprøjtedispenser, Microlab® 600
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
- Dispenserer volumener fra 100 nl til 50 ml
- Stabil væskevej, der kan anvendes til barske kemikalier
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
Certifications: CE marked. CSA certified.
Accessories information: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Ordering information: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
- Beskrivelse:Enkelt sprøjtedispenser med avanceret controller, Microlab 620-DIS, en 1 ml BFP standardsprøjte
- Seq:02
Accessories
- Beskrivelse
- Beskrivelse
- HAMI1171-02Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEP1 * 1 Items - 648,00 kr
- Beskrivelse
- HAMI1173-02Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEP1 * 1 Items - 648,00 kr
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- HAMI61500-05New Microlab 600 controller, includes SD card slot, serial port, large and small USB port1 * 1 Items - 24.320,00 kr
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- HAMI61500-03Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)1 * 1 Items - 2.590,00 kr
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- 613-1585Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP1 * 1 Items - 488,00 kr
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- HAMI61500-02Basic to Advanced Controller upgrade kit, including 2 GB SD card1 * 1 Items - 6.440,00 kr