Orders LinkContactUsLinkComponent
Accessories for columns INdEX™
Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
Katalog # 18-0005-45
Leverandør:   Cytiva
Accessories for columns INdEX™
Katalog # 18-0005-45
Leverandør:  Cytiva
Accessories for columns INdEX™
Accessories for columns INdEX™
Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
Price
3.400,00 kr
Log ind for tilgængelighed
1 * 1 Items
Specifikationer
  • Beskrivelse:
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
  • Seq:
    17