Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant MAGE3 (from E. coli)

Human Recombinant MAGE3 (from E. coli)

Katalog # ABCAAB99958-250
Marketsource
Leverandør:   Abcam
Human Recombinant MAGE3 (from E. coli)
Katalog # ABCAAB99958-250
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant MAGE3 (from E. coli)
Human Recombinant MAGE3 (from E. coli)
CAS nummer:  
Price
17.590,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human MAGE3 protein is a Human Full Length protein, in the 1 to 314 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    MAGE3
  • Protein synonyms:
    MAGE3|MAGEA3|Melanoma-associated antigen 3|Antigen MZ2-D|Cancer/testis antigen 1.3|MAGE-3 antigen|CT1.3
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB99958-250
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Renhed:
    >90%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer