Human Recombinant LC3B (from E. coli)
Catalog # ABCAAB103506-500
Marketsource
Supplier: Abcam
Human Recombinant LC3B (from E. coli)
Catalog # ABCAAB103506-500
Supplier: Abcam
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Recombinant Human LC3B protein is a Human Full Length protein, in the 1 to 140 aa range, expressed in E. coli, with >95% purity and suitable for SDS-PAGE, MS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Konjugation:Unconjugated
- Form:Liquid
- Protein/peptide name:LC3B
- Protein synonyms:MAP1ALC3|MAP1LC3B|Microtubule-associated proteins 1A/1B light chain 3B|Autophagy-related protein LC3 B|Autophagy-related ubiquitin-like modifier LC3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B light chain 3 B|Microtubule-associated protein 1 light chain 3 beta|MAP1A/MAP1B LC3 B
- Biologisk aktivitet:Standard
- Koncentration:0,5 mg/ml
- Dyrefri:N
- Shipping temperature:20
- Cat. No.:ABCAAB103506-500
- Protein/peptide type:Recombinant
- Formulering:pH: 8. Constituents: 20% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol.
- Renhed:>95%
- Tested applications:MS, SDS-PAGE
- Kilde:E. coli
- Species:Human
- Opbevaring betingelser:Store at −20 °C



