Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant PGLS (from E. coli)

Human Recombinant PGLS (from E. coli)

Katalog # ABCAAB101059-250
Marketsource
Leverandør:   Abcam
Human Recombinant PGLS (from E. coli)
Katalog # ABCAAB101059-250
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant PGLS (from E. coli)
Human Recombinant PGLS (from E. coli)
CAS nummer:  
Price
12.540,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human PGLS protein is a Human Full Length protein, in the 1 to 258 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, ELISA.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    PGLS
  • Protein synonyms:
    6-phosphogluconolactonase|6PGL
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    1 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB101059-250
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,58% Sodium chloride, 0,316% Tris HCl, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Renhed:
    >95%
  • Tested applications:
    ELISA, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer