Transport container with screw cap
60 ml, 20 mm, 126 mm, with insert/print, 300/300 (bag/ctn)
Katalog # BOET08.023.0600
Leverandør: BOETTGER PAUL
Gå til:
Product Details & Documents
Made of PP, translucent. Mailing containers for medical and biohazard samples.
- Optimal protection from external elements through cylindrical design
- Absorbent paper insert provides additional protection, easy to change, multiple-use very simple
- Biohazard and recycling symbol clearly visible
Mailing containers Ø 30 mm fits into current mailboxes with 35 mm-wide slot, containers 44/114 mm suitable for mailing common culture mediums and containers 30/126 mm with cap suitable for Port-a-Cul.
Certifications: Corresponding to DIN EN 829 (with insert/print).
Bestillingsoplysninger: Transport tubes and screw caps have to be ordered separate.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Udv.Ø:20 mm
- Pakket:300/300 (bag/ctn)
- Version:with insert/print
- Kapacitet:60 ml
- Højde:126 mm
- Seq:06



