Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Multi-parameter meter, handheld, SevenGo Duo pro™ SG98

Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (1,8 m cable)
Katalog # 665-0238
Leverandør:   Mettler - Toledo
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
Katalog # 665-0238
Leverandør:  Mettler - Toledo
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98

Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (1,8 m cable)

Price
29.950,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (optical RDO® technology). Ideal for special analyses, particularly in environmental, chemical and food and beverage industry.

  • Intuitive, user friendly operation using menu guidance in ten languages and backlit display
  • Fast data transmission thanks to wireless IR communication interface
  • Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
  • Manual or automatic air pressure compensation for reliable results
  • Optical oxygen sensor based on RDO® technology; reliable 'Intelligent Sensor Management' (ISM®)
  • Rugged watertight housing (IP 67)

Electrode Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m cable); Electrode Kit with SG98 also includes InLab® Expert Pro ISM®.

Field Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m or 5 m cable), compact field case, rubber holster and OptiOx™ protective guard; Field Kit with SG98 also includes InLab® Expert Pro ISM®.

 : Meter only is supplied without probes.
The Electrode Kit includes the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 m cable).
The Field Kits include the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 or 5 m cable), OptiOx protective guard, rubber holster, calibration sachets, sample bottles and Field Compact Case.

Specifikationer
  • Beskrivelse:
    Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (1,8 m cable)
  • mV - Område:
    ±1999,9
  • D.O. Nøjagtighed:
    ±0,1 mg/l (0 - 8)
    ±0,2 mg/l (8 - 20)
    ±10% (20 - 50)
  • ISE Område:
    1,00-9 - 9,99+9
  • Datalog Kapacitet:
    500 measurements
  • ISE Enheder:
    mg/l, mmol/l, mol/l
  • pH - Kalibrering:
    Max. 5 points, 7 predefined buffer groups
  • Tilslutninger:
    IR to printer or PC via RS232 or USB
  • ISE Nøjagtighed:
    ±0,5%
  • D.O. - Aflæsningsinterval:
    0,01 mg/L
  • D.O. - mætning:
    0 - 500%
  • Output:
    DO probe: Mini LTW
    Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)
  • Vægt:
    0,368 kg
  • mV - Aflæsningsinterval:
    0,1
  • mV - Nøjagtighed:
    ±0,1
  • pH - Aflæsningsinterval:
    0,1/0,01/0,001
  • Strømforsyning:
    4×1,5 V AA batteries or 1,3 V NiMH accumulators
  • Overensstemmelse:
    IP 67/GLP
  • B×D×H:
    90×45×220 mm
  • pH - Nøjagtighed:
    ±0,002
  • Temperaturinterval:
    DO: 0...+50 °C
    pH (ATC): –5...+130 °C
    MTC: −30...+130 °C
  • pH - Område:
    –2,000...+19,999
  • DO Koncentration:
    0,00 - 50,00 mg/L
  • Temperature accuracy:
    DO: ±0,1 / pH: ±0,2 °C
  • Cat. No.:
    665-0238
  • Model:
    SG98
  • Display:
    Dot matrix LCD
Accessories

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0076
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 14.150,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ beskyttelsesskærm
    OptiOx™ beskyttelsesskærm
    664-0073
    Log ind for tilgængelighed
    1 * 1 Items - 4.570,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ kalibreringsrør
    OptiOx™ kalibreringsrør
    664-0072
    Log ind for tilgængelighed
    1 * 1 Items - 2.480,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • RDO udskiftningslåg til InLab® OptiOx™
    RDO udskiftningslåg til InLab® OptiOx™
    664-0078
    Log ind for tilgængelighed
    1 * 1 Items - 2.390,00 kr

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0077
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 16.210,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ BOD adapter
    OptiOx™ BOD adapter
    664-0074
    Log ind for tilgængelighed
    1 * 1 Items - 1.340,00 kr

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0075
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 14.780,00 kr