Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98

Multi-parameter meter, handheld, SevenGo Duo pro™ SG98

Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (5 m cable)
Katalog # 665-0239
Leverandør:   Mettler - Toledo
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
Katalog # 665-0239
Leverandør:  Mettler - Toledo
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98

Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (5 m cable)

Price
33.400,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (optical RDO® technology). Ideal for special analyses, particularly in environmental, chemical and food and beverage industry.

  • Intuitive, user friendly operation using menu guidance in ten languages and backlit display
  • Fast data transmission thanks to wireless IR communication interface
  • Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID
  • Manual or automatic air pressure compensation for reliable results
  • Optical oxygen sensor based on RDO® technology; reliable 'Intelligent Sensor Management' (ISM®)
  • Rugged watertight housing (IP 67)

Electrode Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m cable); Electrode Kit with SG98 also includes InLab® Expert Pro ISM®.

Field Kit with SG9 includes InLab® OptiOx™ probe (IP 67, 1,8 m or 5 m cable), compact field case, rubber holster and OptiOx™ protective guard; Field Kit with SG98 also includes InLab® Expert Pro ISM®.

Leveringsinformation: Meter only is supplied without probes.
The Electrode Kit includes the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 m cable).
The Field Kits include the combined pH electrode InLab® Expert Pro-ISM and the optical DO sensor InLab® OptiOx (both IP 67, 1,8 or 5 m cable), OptiOx protective guard, rubber holster, calibration sachets, sample bottles and Field Compact Case.

Specifikationer
  • Beskrivelse:
    Handheld multi-parameter meter, SevenGo Duo pro™ SG98, Field Kit (5 m cable)
  • mV - Område:
    ±1999,9
  • D.O. Nøjagtighed:
    ±0,1 mg/l (0 - 8)
    ±0,2 mg/l (8 - 20)
    ±10% (20 - 50)
  • ISE Område:
    1,00-9 - 9,99+9
  • Datalog Kapacitet:
    500 measurements
  • ISE Enheder:
    mg/l, mmol/l, mol/l
  • pH - Kalibrering:
    Max. 5 points, 7 predefined buffer groups
  • Tilslutninger:
    IR to printer or PC via RS232 or USB
  • ISE Nøjagtighed:
    ±0,5%
  • D.O. - Aflæsningsinterval:
    0,01 mg/L
  • D.O. - mætning:
    0 - 500%
  • Output:
    DO probe: Mini LTW
    Other sensors: BNC (<1012 Ohm); RCA (Cinch) (all IP 67)
  • Vægt:
    0,368 kg
  • mV - Aflæsningsinterval:
    0,1
  • mV - Nøjagtighed:
    ±0,1
  • pH - Aflæsningsinterval:
    0,1/0,01/0,001
  • Strømforsyning:
    4×1,5 V AA batteries or 1,3 V NiMH accumulators
  • Overensstemmelse:
    IP 67/GLP
  • B×D×H:
    90×45×220 mm
  • pH - Nøjagtighed:
    ±0,002
  • Temperaturinterval:
    DO: 0...+50 °C
    pH (ATC): –5...+130 °C
    MTC: −30...+130 °C
  • pH - Område:
    –2,000...+19,999
  • DO Koncentration:
    0,00 - 50,00 mg/L
  • Temperature accuracy:
    DO: ±0,1 / pH: ±0,2 °C
  • Cat. No.:
    665-0239
  • Model:
    SG98
  • Display:
    Dot matrix LCD
Accessories

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0076
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 14.150,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ beskyttelsesskærm
    OptiOx™ beskyttelsesskærm
    664-0073
    Log ind for tilgængelighed
    1 * 1 Items - 4.570,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ kalibreringsrør
    OptiOx™ kalibreringsrør
    664-0072
    Log ind for tilgængelighed
    1 * 1 Items - 2.480,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • RDO udskiftningslåg til InLab® OptiOx™
    RDO udskiftningslåg til InLab® OptiOx™
    664-0078
    Log ind for tilgængelighed
    1 * 1 Items - 2.390,00 kr

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0077
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 16.210,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • OptiOx™ BOD adapter
    OptiOx™ BOD adapter
    664-0074
    Log ind for tilgængelighed
    1 * 1 Items - 1.340,00 kr

Produktinformation

  • Katalog #
    Model
    Koncentration
    Tilgængelighed
    Price
  • Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    Optical DO probe InLab® OptiOx™, with temperature sensor, IP 67
    664-0075
    InLab® OptiOx™
    0 - 50 mg/l (0 - 500%)
    Log ind for tilgængelighed
    1 * 1 Items - 14.780,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.