Accessories for VACUSIP
Tubing set consist of VACUSIP hand operator, silicone tubing (1,2 m and 0,22 m) and non sterile filter (0,45 µm)
Catalog # 181-0260
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Tubing set consist of VACUSIP hand operator, silicone tubing (1,2 m and 0,22 m) and non sterile filter (0,45 µm)
- Seq:15