MITOCHONDRIAL OXPHOS (HUMAN) ANTIBODY CO 1 * 1 Each
Katalog # NOVGACMT001-360UG
Leverandør: Merck Millipore (Novagen)
MITOCHONDRIAL OXPHOS (HUMAN) ANTIBODY CO 1 * 1 Each
Katalog # NOVGACMT001-360UG
Leverandør: Merck Millipore (Novagen)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
Sealing film for manual application, Parafilm® M
Leverandør: HEATHROW
Handsker, nitril, Kimtech™ Purple Nitrile™
Leverandør: KIMBERLY CLARK
VWR®, Serologiske pipetter, Standard type
Leverandør: VWR Collection
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
VWR®, Ansigtsservietter
Leverandør: VWR Collection
pH-teststrimler, MQuant®, Supelco®
Leverandør: Merck
Vand, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Leverandør: VWR Collection
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection