Orders LinkContactUsLinkComponent
Objectives for BA series
Objectives for BA series
EC-H PL objective (negative phase) (spring), Ph 40×/0,65
Katalog # MOTI1101001703411
Leverandør:   MOTIC
Objectives for BA series
Katalog # MOTI1101001703411
Leverandør:  MOTIC
Objectives for BA series
Objectives for BA series
EC-H PL objective (negative phase) (spring), Ph 40×/0,65
Price
2.840,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Forstørrelse:
    Ph 40×/0,65
  • Beskrivelse:
    EC-H PL objective (negative phase) (spring)
  • Seq:
    213