Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumchlorid 0,9%, Ecotainer®

Natriumchlorid 0,9%, Ecotainer®

Bottles of 500 ml, 10
Katalog # BRAU3570130
Leverandør:   B. Braun
Natriumchlorid 0,9%, Ecotainer®
Katalog # BRAU3570130
Leverandør:  B. Braun
CAS nummer:  7647-14-5
Natriumchlorid 0,9%, Ecotainer®
Natriumchlorid 0,9%, Ecotainer®

Bottles of 500 ml, 10

CAS nummer:  7647-14-5
Price
256,00 kr
Log ind for tilgængelighed
1 * 10 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Sterile and pyrogen-free topical rinse solution.

  • PVC- and latex-free
  • Sterile and non-pyrogenic
  • Handy container with screw cap

Ecotainer® has been designed for intra- and post-operative irrigation.

Synonymer:  Saline, NaCl, Sea salt, Halite
  • CAS nummer:7647-14-5
  • Formel:NaCl
  • MDL nummer:MFCD00003477
  • Molecular Weight:58.44 g/mol

Documents

Specifikationer

Specifikationer

  • Pakn:
    10
  • Pk Info:
    Bottles of 500 ml

Specifikationer