Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carboxyl coupling resin

Carboxyl coupling resin

Carboxyl coupling resin (Immobilized DADPA (Diaminodipropylamine)), 25 mL
Katalog # 786-797
Leverandør:   G-Biosciences
Carboxyl coupling resin
Katalog # 786-797
Leverandør:  G-Biosciences
CAS nummer:  
Carboxyl coupling resin
Carboxyl coupling resin

Carboxyl coupling resin (Immobilized DADPA (Diaminodipropylamine)), 25 mL

CAS nummer:  
Price
2.140,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents

Carboxyl coupling resin consists of 6% cross-linked agarose with covalently linked diaminodipropylamine (DADPA) to generate a free primary amine at the end of a long spacer arm. Molecules, including proteins, peptides and nucleic acids, are covalently coupled to the free primary amines, and the stable columns are ideal for affinity purification of antibodies and other interacting partners.

Specifikationer

Specifikationer

  • Pakn:
    25 mL
  • Beskrivelse:
    Carboxyl coupling resin (Immobilized DADPA (Diaminodipropylamine))

Specifikationer