Orders LinkContactUsLinkComponent
Biotin kits, HOOK™
Biotin kits, HOOK™
NHS-LC-LC-Biotin kit, 1 KIT
Katalog # BS-03
Leverandør:   G-Biosciences
Biotin kits, HOOK™
Katalog # BS-03
Leverandør:  G-Biosciences
CAS nummer:  
Biotin kits, HOOK™
Biotin kits, HOOK™
NHS-LC-LC-Biotin kit, 1 KIT
CAS nummer:  
Price
3.760,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
HOOK™ Biotin Reagent that conjugates to proteins through amines, sulfhydryls, carboxyls or carbohydrates. The amine and sulfhydryl coupling HOOK™ Biotin Reagents couple directly to the protein through their reactive groups, however the carboxyl coupling HOOK™ Biotin Reagents require a carbodiimide crosslinker and the carbohydrate coupling HOOK™ Biotin Reagents require carbohydrate oxidation before coupling.
  • Optimizer Buffer™ for improved coupling efficiency
  • SpinOUT™ gel filtration columns for rapid (<10 minutes) purification
  • Biotin assay reagents to determine level of biotin incorporation
  • Labels 1 to 10 mg protein/reaction
  • Suitable for 10 coupling reactions
The kits include EDC as the carbodiimide crosslinker in the carboxyl coupling kits and sodium meta-periodate for carbohydrate oxidation in the carbohydrate coupling kits. Kit also contains a specific Optimizer Buffer™ that provides the optimal reaction conditions for each HOOK™ Biotin Reagent.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    NHS-LC-LC-Biotin kit
Specifikationer
Accessories
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-V, 5X
    Optimizer Buffer™-V, 5X
    BKC-08
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-III, 5X
    Optimizer Buffer™-III, 5X
    BKC-06
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-IV, 5X
    Optimizer Buffer™-IV, 5X
    BKC-07
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-VI, 5X
    Optimizer Buffer™-VI, 5X
    BKC-09
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-II,5X
    Optimizer Buffer™-II,5X
    BKC-05
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-I, 5X
    Optimizer Buffer™-I, 5X
    BKC-04
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr