Orders LinkContactUsLinkComponent
Nutrient Pad Sets, NKS e.motion
Food/ beverage, 0,45 µm, Yeast and moulds, 100
Katalog # SART14058-50-RKN
Leverandør:   Sartorius
Nutrient Pad Sets, NKS e.motion
Katalog # SART14058-50-RKN
Leverandør:  Sartorius
CAS nummer:  
Nutrient Pad Sets, NKS e.motion
Nutrient Pad Sets, NKS e.motion
Food/ beverage, 0,45 µm, Yeast and moulds, 100
CAS nummer:  
Price
1.990,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Nutrient pad sets for colony counting by the membrane filter method, specially designed for the Microsart e.motion dispenser. These nutrient pad consist of dehydrated culture media inserted into pre-sterilised Petri dishes. This alternative to agar media has a long storage life of 24 months at room temperature.
  • Economical, time-saving microbiological control
  • High quality and reproducible results for up to 24 months
  • Compliant with international standards and references
  • Easy handling
Specifikationer
Specifikationer
  • Pakn:
    100
  • Porestørrelse:
    0,45 µm
  • Model:
    Food/ beverage
  • Til:
    Yeast and moulds
Specifikationer