Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
HPLC columns, Syncronis™ HILIC

HPLC columns, Syncronis™ HILIC

Beskyttelsespatron, HILIC, 100 Å, 5 µm, 4,00 mm, 10 mm
Katalog # HYPE97505-014001
Leverandør:   Thermo Fisher Scientific
HPLC columns, Syncronis™ HILIC
Katalog # HYPE97505-014001
HPLC columns, Syncronis™ HILIC
HPLC columns, Syncronis™ HILIC

Beskyttelsespatron, HILIC, 100 Å, 5 µm, 4,00 mm, 10 mm

Price
5.130,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents

The Hydrophilic Interaction Liquid Chromatography (HILIC) technique offers complementary selectivity to reversed phase. With the ability to retain highly polar and hydrophilic compounds, these columns have been developed to aid the analysis of compounds that are traditionally difficult to retain using conventional C18 columns.

  • Zwitterion ligand gives neutral, highly polar surface
  • No need for ion-pair reagents or derivatisation
  • Highly pure, high surface area silica

Furthermore, the highly organic mobile phase contains low salt levels, making Syncronis™ HILIC columns ideal for use with electrospray mass spectroscopy.

Surface area: 320 m²/g

Carbon load: 5%

pH range: 2 - 8

Specifikationer
  • Kolonne form:
    Beskyttelsespatron
  • Ø indv.:
    4,00 mm
  • Partikelstørrelse:
    5 µm
  • Porestørrelse:
    100 Å
  • Længde:
    10 mm
  • Fase:
    HILIC
  • Cat. No.:
    HYPE97505-014001
  • Kulstofbelastning:
    5%
  • pH-område:
    2 - 8
  • Surface area:
    320 m²/g
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.