HPLC columns, Syncronis™ HILIC
Analytisk, HILIC, 100 Å, 5 µm, 2,10 mm, 50 mm
HYPE97505-052130
The Hydrophilic Interaction Liquid Chromatography (HILIC) technique offers complementary selectivity to reversed phase. With the ability to retain highly polar and hydrophilic compounds, these columns have been developed to aid the analysis of compounds that are traditionally difficult to retain using conventional C18 columns.
- Zwitterion ligand gives neutral, highly polar surface
- No need for ion-pair reagents or derivatisation
- Highly pure, high surface area silica
Furthermore, the highly organic mobile phase contains low salt levels, making Syncronis™ HILIC columns ideal for use with electrospray mass spectroscopy.
Surface area: 320 m²/g
Carbon load: 5%
pH range: 2 - 8
- Kolonne form:Analytisk
- Ø indv.:2,10 mm
- Partikelstørrelse:5 µm
- Porestørrelse:100 Å
- Længde:50 mm
- Fase:HILIC
- Cat. No.:HYPE97505-052130
- Kulstofbelastning:5%
- pH-område:2 - 8
- Surface area:320 m²/g



