Product Details & Documents
PerfeCTa® Multiplex qPCR SuperMix, low ROX is a 2X concentrated, ready to use reaction cocktail that contains all components, except primers, probe(s), and template for real time qPCR on Applied Biosystems 7500 or Stratagene MX series of real time PCR systems.
PerfeCTa® Multiplex qPCR SuperMix, low ROX delivers dynamic range and sensitivity to multiplexed qPCR that is comparable to that for singleplex qPCR probe assays without the need for limiting or variable primer concentrations.
Delivery information: Each kit contains 2X reaction buffer containing optimised concentrations of MgCl₂, dNTPs (dATP, dCTP, dGTP, dTTP), AccuStart™ Taq DNA polymerase, ROX reference dye (for 580 to 585 nm excitation), and stabilisers.
Documents
Specifikationer
Specifikationer
- Cycler kompatibilitet:ABI 7500/7500 Fast/QuantStudio™, Agilent AriaMx, Douglas Scientific IntelliQube®, Stratagene Mx
- Pakn:200 Assays
- Beskrivelse:PerfeCTa® Multiplex qPCR SuperMix, low ROX
- Multiplexing:Yes
- Cat. No.:733-2088
- Ansøgning:qPCR probe