Wound cleansers
Salvequick cleaning wipes refill box, incl. 20 pcs sterile wound cleansers (110×120 mm)
Katalog # 118-0250
Leverandør: Orkla Care AB / Cederroth
Gå til:
Product Details & Documents
Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (0,9% NaCl), are intended for the mechanical cleansing of minor wounds when clean tap water is not available.
- Tissues are packed in sealed sachets, which prevent leakage and guarantee sterility
- Savett sachets are packed in a refill box
- Salvequick cleansers fit into first-aid-stations
Documents
VWR® for Lab Safety
Produktlitteratur
Specifikationer
- Beskrivelse:Salvequick cleaning wipes refill box, incl. 20 pcs sterile wound cleansers (110×120 mm)