Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
PIG® poly spill containment pallets

PIG® poly spill containment pallets

2-drum poly spill containment pallet, 250 L, 1350×730×410 mm
Katalog # 121-0183
Leverandør:   NEW PIG
PIG® poly spill containment pallets
Katalog # 121-0183
Leverandør:  NEW PIG
PIG® poly spill containment pallets
PIG® poly spill containment pallets

2-drum poly spill containment pallet, 250 L, 1350×730×410 mm

Price
3.130,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These economical pallets are strong enough to handle long-term storage of fully loaded steel drums.

  • Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
  • Molded-in sump catches leaks, drips and spills to help you comply with regulations and keep your storage areas clean and safe
  • Strong enough to support four fully loaded steel or poly drums
  • Textured grating adds traction and lifts out for easy sump access
  • 2-way forklift entry allows easy movement from either direction (unloaded only)

Nestable design permits stacking when not in use; lowers cost of bulk shipping.

Specifikationer
  • Kapacitet:
    250 L
  • Beskrivelse:
    2-drum poly spill containment pallet
  • B×D×H:
    1350×730×410 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.