Orders LinkContactUsLinkComponent
Sample tubes
Sample tubes
11 ml, 108 mm, 16 mm, PP
Katalog # METR6.2743.057
Leverandør:   METROHM
Sample tubes
Katalog # METR6.2743.057
Leverandør:  METROHM
Sample tubes
11 ml, 108 mm, 16 mm, PP
Price
3.010,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
Ideal for IC sample processors and VA autosamplers.
Specifikationer
  • Udv.Ø:
    16 mm
  • Kapacitet:
    11 ml
  • Længde:
    108 mm
  • Materiale:
    PP
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Perforated stopper, PP, for 2,5 and 11 ml sample tubes
    Perforated stopper, PP, for 2,5 and 11 ml sample tubes
    METR6.2743.077
    Log ind for tilgængelighed
    1 * 200 Items - 847,00 kr