Drying trays
500×300×40 mm
Catalog # JUCHVLSOBAYB807861
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Made of acid resistant stainless steel.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for easy cleaning
Specifications
- Seq:04
- B×D×H:500×300×40 mm