Drying trays
400×300×40 mm
JUCHVLSOBAYB807841
Made of acid resistant stainless steel.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for easy cleaning
- Seq:03
- B×D×H:400×300×40 mm