Drying trays
180×130×40 mm
Katalog # JUCHVLSOBAYB807801
Leverandør: JUCHHEIM Laborgerate GmbH
Gå til:
Product Details & Documents
Made of acid resistant stainless steel.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for easy cleaning
Specifikationer
- Seq:01
- B×D×H:180×130×40 mm