Double bossheads, cross shape
Double bosshead, cross shape, with M8 stainless steel thumb bolt, Jern, 16 mm
Catalog # JUCHVL04850S
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Bossheads made from either epoxy coated malleable iron or from stainless steel.
Specifications
- Beskrivelse:Double bosshead, cross shape, with M8 stainless steel thumb bolt
- Materiale:Jern
- Til stang Ø:16 mm