Double bosshead
Double bosshead with M8 brass thumb bolt, 16 mm
Katalog # JUCHVL04810
Leverandør: JUCHHEIM Laborgerate GmbH
Gå til:
Product Details & Documents
Double bosshead made from epoxy coated malleable iron.
Certifications: DIN 12895
Specifikationer
- Beskrivelse:Double bosshead with M8 brass thumb bolt
- Seq:01
- Til stang Ø:16 mm