HPLC columns, VariTide RPC
VariTide RPC bulk media, 100 g, 6 µm
Katalog # VARIPL1412-4A05
Leverandør: VARIAN
Gå til:
Product Details & Documents
VariTide RPC column is designed for use across the pH range, making it suitable for the analysis and purification of acid or basic and hydrophilic or hydrophobic peptides, using acid, neutral and basic eluents.
- Separations run at elevated temperature and pH, and under acidic, neutral and basic conditions
- Robustness and durability
- Single, easy to use, universal column
- A single column to cover the full range of synthetic peptides
A high efficiency, small particle packing, with a pore size optimised for high capacity across the peptide size range, from 5 to 60 amino acids.
Specifikationer
- Partikelstørrelse:6 µm
- Beskrivelse:VariTide RPC bulk media, 100 g
- Seq:04



