Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 13,000 Da., 1 g, 1
Katalog # VARIPL2023-0001
Leverandør: VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Katalog # VARIPL2023-0001
Leverandør: VARIAN
CAS nummer:
Gå til:
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
- Many solvent options increase applicability
- Stringent quality control improves performance
- Proprietary manufacturing methods ensure consistent supply
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,03, nominal Mp 13,000 Da.
- Pk Info:1 g