Polymethylmethacrylate standards, GPC/SEC columns calibration
Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da., 5 g, 1
Catalog # VARIPL2022-2005
Supplier: VARIAN
Polymethylmethacrylate standards, GPC/SEC columns calibration
Catalog # VARIPL2022-2005
Supplier: VARIAN
CAS Number:
Jump to:
Product Details & Documents
Polymethylmethacrylate (PMMA) standards are extremely versatile as they can be used for organic GPC with a wide range of medium polarity eluents, such as tetrahydrofuran, toluene, methyl ethyl ketone and ethyl acetate.
- Many solvent options increase applicability
- Stringent quality control improves performance
- Proprietary manufacturing methods ensure consistent supply
Specifications
Specifications
- Pakn:1
- Beskrivelse:Polymethylmethacrylate, individual polymer molecular weights, nominal Mw/Mn 1,19, nominal Mp 500 Da.
- Pk Info:5 g