Polystyrene standards, GPC/SEC columns calibration
Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,01, quantities 01 g, 1
Katalog # VARIPL2013-6001
Leverandør: VARIAN
Polystyrene standards, GPC/SEC columns calibration
Katalog # VARIPL2013-6001
Leverandør: VARIAN
CAS nummer:
Gå til:
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
- Compatible with most organic solvents
- Certificate of analysis meets international protocols
- Calibration capability for virtually all applications
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:1
- Beskrivelse:Polystyrene, individual polymer molecular weight, nominal Mw/Mn 1,01, quantities 01 g