Orders LinkContactUsLinkComponent
Pre-prepared polystyrene kits, EasiCal™
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-2, 580 - 400000 g/mol, 5
Katalog # VARIPL2010-0605
Leverandør:   VARIAN
Pre-prepared polystyrene kits, EasiCal™
Katalog # VARIPL2010-0605
Leverandør:  VARIAN
CAS nummer:  
Pre-prepared polystyrene kits, EasiCal™
Pre-prepared polystyrene kits, EasiCal™
Polystyrene PS-2, 580 - 400000 g/mol, 5
CAS nummer:  
Price
16.620,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
  • Easy three step process with no fuss
  • Cost effective
  • Only two injections for improved productivity
  • Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.
Specifikationer
Specifikationer
  • Pakn:
    5
  • Beskrivelse:
    Polystyrene PS-2, 580 - 400000 g/mol