Orders LinkContactUsLinkComponent
Polystyrene standards, GPC/SEC columns calibration
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-L2-10, 10×0,5 g, 1
Katalog # VARIPL2010-0105
Leverandør:   VARIAN
Polystyrene standards, GPC/SEC columns calibration
Katalog # VARIPL2010-0105
Leverandør:  VARIAN
CAS nummer:  
Polystyrene standards, GPC/SEC columns calibration
Polystyrene calibration kit, S-L2-10, 10×0,5 g, 1
CAS nummer:  
Price
9.600,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
  • Compatible with most organic solvents
  • Certificate of analysis meets international protocols
  • Calibration capability for virtually all applications
Documents
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Polystyrene calibration kit, S-L2-10, 10×0,5 g