High efficiency purification columns, biomolecule seperation
Seperation of synthetic peptides & recombinant peptides and proteins by reverse phase, PLRP-S, 300 Å, 8 µm, 100,00 mm, 300 mm
Catalog # VARIPL1812-6801
Supplier: VARIAN
High efficiency purification columns, biomolecule seperation
Catalog # VARIPL1812-6801
Supplier: VARIAN
Jump to:
Product Details & Documents
Specifications
- Ø indv.:100,00 mm
- Partikelstørrelse:8 µm
- Porestørrelse:300 Å
- Beskrivelse:Seperation of synthetic peptides & recombinant peptides and proteins by reverse phase
- Længde:300 mm
- Fase:PLRP-S
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



