High efficiency purification columns, biomolecule seperation
Seperation of synthetic peptides & recombinant peptides and proteins by reverse phase, PLRP-S, 100 Å, 8 µm, 100,00 mm, 300 mm
Katalog # VARIPL1812-6800
Leverandør: VARIAN
High efficiency purification columns, biomolecule seperation
Katalog # VARIPL1812-6800
Leverandør: VARIAN
Gå til:
Product Details & Documents
Specifikationer
- Ø indv.:100,00 mm
- Partikelstørrelse:8 µm
- Porestørrelse:100 Å
- Beskrivelse:Seperation of synthetic peptides & recombinant peptides and proteins by reverse phase
- Længde:300 mm
- Fase:PLRP-S



