Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Columns and standards for GPC/SEC

Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 50, 8 µm, 7,50 mm, 300 mm
  VARIPL1149-6850
 :   VARIAN
Columns and standards for GPC/SEC
  VARIPL1149-6850
 :  VARIAN
Columns and standards for GPC/SEC
Columns and standards for GPC/SEC

Aqueous SEC of polymers, PL-aquagel-OH 50, 8 µm, 7,50 mm, 300 mm

 
22.840,00 kr
   
1 * 1 Items
 
 
  • Ø indv.:
    7,50 mm
  • Partikelstørrelse:
    8 µm
  • Beskrivelse:
    Aqueous SEC of polymers, PL-aquagel-OH 50
  • Længde:
    300 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.