High efficiency purification columns, biomolecule seperation
Seperation of synthetic peptides & recombinant peptides and proteins by anion exchange, PLRP-S, 1000 Å, 8 µm, 7,50 mm, 300 mm
Katalog # VARIPL1112-6802
Leverandør: VARIAN
High efficiency purification columns, biomolecule seperation
Katalog # VARIPL1112-6802
Leverandør: VARIAN
Gå til:
Product Details & Documents
Specifikationer
- Ø indv.:7,50 mm
- Partikelstørrelse:8 µm
- Porestørrelse:1000 Å
- Beskrivelse:Seperation of synthetic peptides & recombinant peptides and proteins by anion exchange
- Længde:300 mm
- Fase:PLRP-S
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



