Aluminium headspace caps
Aluminium cap, ridged, Butyl grey stopper
Katalog # PERCN9303350
Leverandør: PerkinElmer
Gå til:
Product Details & Documents
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
- Temperature resistance up to 130 °C
- Very economical and has low resistance to coring
Specifikationer
- Septum:Butyl grey stopper
- Beskrivelse:Aluminium cap, ridged
- Seq:01