Orders LinkContactUsLinkComponent
Aluminium headspace caps
Aluminium headspace caps
Aluminium cap, ridged, Butyl grey stopper
Katalog # PERCN9303350
Leverandør:   PerkinElmer
Aluminium headspace caps
Katalog # PERCN9303350
Leverandør:  PerkinElmer
Aluminium headspace caps
Aluminium cap, ridged, Butyl grey stopper
Price
6.170,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
  • Temperature resistance up to 130 °C
  • Very economical and has low resistance to coring
Specifikationer
  • Septum:
    Butyl grey stopper
  • Beskrivelse:
    Aluminium cap, ridged
  • Seq:
    01