Orders LinkContactUsLinkComponent
SPE cartridges, SPEC disk
SPE cartridges, SPEC disk
Straight barrel cartridge, MP1, 3 ml, 15 mg
Katalog # VARIA5311120B
Leverandør:   VARIAN
SPE cartridges, SPEC disk
Katalog # VARIA5311120B
Leverandør:  VARIAN
SPE cartridges, SPEC disk
Straight barrel cartridge, MP1, 3 ml, 15 mg
Price
30.060,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
SPEC Disk SPE delivers superior flow characteristics and trouble free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
Bond Elut™ SPEC is a monolithic sorbent bed manufactured with ultraclean polypropylene plastic and bonded-silica impregnated on a glass fiber disc, resulting in an SPE product that delivers faster sample processing and lower costs. SPEC can result in increased productivity and lower extraction costs, without sacrificing the quality of the analytical results. For some applications, the elution volume is very small, meaning that the need to dry down and reconstitute the extract is eliminated. Integrated filtration protects against clogging.
Specifikationer
  • Kapacitet:
    3 ml
  • Beskrivelse:
    Straight barrel cartridge
  • Seq:
    01
  • Lejevægt:
    15 mg
  • Fase:
    MP1