Orders LinkContactUsLinkComponent
Chromatography paper
Chromatography paper
Chromatography paper (SA), 114×305 mm
Katalog # VARIA120B12
Leverandør:   VARIAN
Chromatography paper
Katalog # VARIA120B12
Leverandør:  VARIAN
Chromatography paper
Chromatography paper
Chromatography paper (SA), 114×305 mm
Price
7.170,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity, QC testing.
  • The porous paper is made of glass microfibers impregnated with silica gel
  • No interference from organic binders
  • Separates lipids and other non-polar compounds
  • Can easily be cut to convenient testing sizes, and can be imprinted
Bestillingsoplysninger: Offers two kinds of paper such as SA (contains sodium salt) and SG (contains potassium salt).
Documents
Specifikationer
  • Beskrivelse:
    Chromatography paper (SA)
  • Størrelse:
    114×305 mm