Orders LinkContactUsLinkComponent
Torches
Axial torches, sheath gas high solids torch with patented injector design, 2,4 mm ID, used with the AGM-1 oxygen
  VARI2010122400
 :   VARIAN
Torches
  VARI2010122400
 :  VARIAN
Torches
Torches
Axial torches, sheath gas high solids torch with patented injector design, 2,4 mm ID, used with the AGM-1 oxygen
10.990,00 kr
1 * 1 Items
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.
  • Beskrivelse:
    Axial torches, sheath gas high solids torch with patented injector design, 2,4 mm ID, used with the AGM-1 oxygen
  • Seq:
    06
Accessories
  •  
    Beskrivelse
  • VARI2010070790
    Spare quartz torch bonnet for radial ICP-OES
    1 * 1 Items - 1.690,00 kr
  •  
    Beskrivelse
  • VARI7210020700
    Torch alignment tool for radial ICP-OES
    1 * 1 Items
  •  
    Beskrivelse
  • VARI7910051290
    GazFit fittings for one-piece and semi-demountable ICP-OES torches
    1 * 1 Items - 1.990,00 kr
  •  
    Beskrivelse
  • VARI7910051590
    Transfer tube FEP for axial ICP
    1 * 1 Items - 1.990,00 kr
  •  
    Beskrivelse
  • VARI7910051490
    Transfer tube FEP Sturman-Masters spray chamber for radial ICP
    1 * 1 Items - 1.990,00 kr
  •  
    Beskrivelse
  • VARI9910107100
    Kit Torch Fittings
    1 * 1 Items - 1.030,00 kr
  •  
    Beskrivelse
  • VARI3610007300
    Pre-optics cone for Liberty Series axial ICP-OES
    1 * 1 Items - 8.550,00 kr
  •  
    Beskrivelse
  • VARI8110002800
    Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OES
    1 * 1 Items - 544,00 kr
  •  
    Beskrivelse
  • VARI2010072900
    Spare semi-demountable quartz torch body for radial ICP-OES
    1 * 1 Items - 11.040,00 kr
  •  
    Beskrivelse
  • VARI2010077500
    Quartz injector tube, 0,8 mm
    1 * 1 Items
  •  
    Beskrivelse
  • VARI2010083400
    Spare semi-demountable quartz torch body for axial ICP-OES
    1 * 1 Items - 11.040,00 kr
  •  
    Beskrivelse
  • VARI9910057000
    Quartz injector tube, 1,4 mm
    1 * 1 Items - 9.570,00 kr
  •  
    Beskrivelse
  • VARI9910057300
    Injector holder kit, includes O-ring
    1 * 1 Items - 1.530,00 kr
  •  
    Beskrivelse
  • VARI9910108200
    O-rings kit
    1 * 1 Items - 314,00 kr