Orders LinkContactUsLinkComponent
Urinanalysis Kit, KOVA®
System Super Pac Kit for 1000 determinations, 1.000 SET
Katalog # 720-2420
Leverandør:   Kova International
Urinanalysis Kit, KOVA®
Katalog # 720-2420
Leverandør:  Kova International
CAS nummer:  
Urinanalysis Kit, KOVA®
Urinanalysis Kit, KOVA®
System Super Pac Kit for 1000 determinations, 1.000 SET
CAS nummer:  
Price
5.770,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Complete urinalysis kit for up to 1000 determinations.
  • 12 ml graduated non-sterile disposable centrifuge tubes manufactured of high-quality, break-resistant clear plastic and marked in 1 ml increments
  • Disposable plastic transfer pipettes that feature a unique lock for one-step, contamination-free decanting, retaining 1 ml of urine after centrifugation
  • Precision manufactured, 10-chambered disposable microscope slides made of optically clear plastic, the Glasstic® slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
 : Contains 100 slides (10 chambered), 1000 KOVA Petters (pipettes with bulge) and 1000 KOVA tubes.
Specifikationer
Specifikationer
  • Pakn:
    1.000 SET
  • Beskrivelse:
    System Super Pac Kit for 1000 determinations
Specifikationer