Urinanalysis Kit, KOVA®
System Super Pac Kit for 1000 determinations, 1.000 SET
Katalog # 720-2420
Leverandør: Kova International
Gå til:
Product Details & Documents
Complete urinalysis kit for up to 1000 determinations.
- 12 ml graduated non-sterile disposable centrifuge tubes manufactured of high-quality, break-resistant clear plastic and marked in 1 ml increments
- Disposable plastic transfer pipettes that feature a unique lock for one-step, contamination-free decanting, retaining 1 ml of urine after centrifugation
- Precision manufactured, 10-chambered disposable microscope slides made of optically clear plastic, the Glasstic® slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
: Contains 100 slides (10 chambered), 1000 KOVA Petters (pipettes with bulge) and 1000 KOVA tubes.
Specifikationer
Specifikationer
- Pakn:1.000 SET
- Beskrivelse:System Super Pac Kit for 1000 determinations