Glove liners, BCR®
Half-finger cleanroom liners, black cuff, Half-finger cleanroom liners, black cuff, L, hvid
Katalog # BERKBGL6.200.LB
Leverandør: BERKSHIRE
Gå til:
Product Details & Documents
These single knit glove liners are made out of high quality 100% continuous filament pre-shrunk nylon. They are lightweight, soft, and comfortable to wear. Available as half-finger and full-finger versions.
- Protect against skin irritation caused by latex or PVC gloves
- Constructed using special seamless techniques
- Ambidextrous for easy stock control
- Launderable for multiple reuse
- Colour-coded cuffs to identify sizes
Suitable for use in cleanrooms.
Specifikationer
- Handske størrelse:L
- Farve:hvid
- Beskrivelse:Half-finger cleanroom liners, black cuff
- Seq:02
- Model:Half-finger cleanroom liners, black cuff
- Cat. No.:BERKBGL6.200.LB
- Ambidextrous/Hånd-specifik:Ambidextrous
- Engangs/Genbrugelig:Genanvendelig