Ceramic beads, SiLibeads®
Ceramic beads, 1,0 -1,20 mm, Zirconiumoxid
LINR9711
Ceramic beads made of Zirconium Oxide / Yttrium stabilised.
- High density
- Very high wear and tear resistance
- Low contamination of the grinding product
- Useable for all modern types of mills and high energy mills (vertical and horizontal)
- Excellent crystal structure avoids bead breakage and reduces the abrasion of mill parts
- No radioactivity
- Conformity to 1935/2004/EC
The exact roundness as well as the smooth and polished surface of the beads contributes to low wear rate.
- Ø:1,0 -1,20 mm
- Beskrivelse:Ceramic beads
- Materiale:Zirconiumoxid