Orders LinkContactUsLinkComponent
Accessories for test sieves
Accessories for test sieves
Sieve pan, 250 mm, 55 mm
Katalog # HAVE205922640
Leverandør:   NEXOPART
Accessories for test sieves
Katalog # HAVE205922640
Leverandør:  NEXOPART
Accessories for test sieves
Accessories for test sieves
Sieve pan, 250 mm, 55 mm
Price
858,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø:
    250 mm
  • Beskrivelse:
    Sieve pan
  • Højde:
    55 mm
  • Seq:
    07