Receiving flasks
3000 ml, Low temperature, KS 35/20
Katalog # BUCH040777
Leverandør: BUCHI
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifikationer
- Top:KS 35/20
- Model:Low temperature
- Kapacitet:3000 ml