Spore strips
Geobacillus stearothermophilus cell line 7953: traditional spore strip 107 (24 month shelf life) / sterilisation process: steam, 30, 6 mm
Katalog # NAMSBS-108
Leverandør: CROSSTEX
Gå til:
Product Details & Documents
Documents
Specifikationer
- Bredde:6 mm
- Beskrivelse:Geobacillus stearothermophilus cell line 7953: traditional spore strip 107 (24 month shelf life) / sterilisation process: steam
- Længde:30
- Seq:03
- Cat. No.:NAMSBS-108